Product Details

SNP ID
rs150210490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31510047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATCTGGGCTCAGCCGAGGGGAA[A/C]CTTAGGGAGGGGACCTGTCGGGGAA
Phenotype
MIM: 606985 MIM: 612323
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELP4 PubMed Links

Gene Details

Gene
ELP4
Gene Name
elongator acetyltransferase complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288725.1 Intron NP_001275654.1
NM_001288726.1 Intron NP_001275655.1
NM_019040.4 Intron NP_061913.3
Gene
IMMP1L
Gene Name
inner mitochondrial membrane peptidase subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304274.1 Intron NP_001291203.1
NM_144981.2 Intron NP_659418.1
XM_005252812.3 Intron XP_005252869.1
XM_011519942.1 Intron XP_011518244.1
XM_011519943.2 Intron XP_011518245.1
XM_011519944.2 Intron XP_011518246.1
XM_011519945.2 Intron XP_011518247.1
XM_011519946.2 Intron XP_011518248.1
XM_011519947.2 Intron XP_011518249.1
XM_017017305.1 Intron XP_016872794.1
XM_017017306.1 Intron XP_016872795.1
XM_017017307.1 Intron XP_016872796.1

View Full Product Details