Product Details
- SNP ID
-
rs11151964
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:74534683 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCAGCCTAATGGATCCCAAACTC[A/G]GGAGAATGGTGAGTAGGACCTCCTC
- Phenotype
-
MIM: 609064
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CNDP1
PubMed Links
Gene Details
- Gene
- CNDP1
- Gene Name
- carnosine dipeptidase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032649.5 |
227 |
Missense Mutation |
AGG,GGG |
R6G |
NP_116038.4 |
View Full Product Details