Product Details

SNP ID
rs11733156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:42398095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGCTTTGCCTTCTCCTTGGCTG[G/T]CTGCGCTGGGGCCCGGCGGGCGCCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105374428 PubMed Links

Gene Details

Gene
LOC105374428
Gene Name
uncharacterized LOC105374428
There are no transcripts associated with this gene.

Gene
SHISA3
Gene Name
shisa family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080505.1 257 Missense Mutation TGG,TGT W13C NP_001073974.1

View Full Product Details