Product Details
- SNP ID
-
rs8068970
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40978962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGTTATCGATCAGACACTGAAT[C/T]TGGGCCAGCTGGGAGCTGTACTGGG
- Phenotype
-
MIM: 616679
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150812789] are located under a probe and SNP(s) [rs8064910] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT40
- Gene Name
- keratin 40
View Full Product Details