Product Details

SNP ID
rs61733264
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:228508727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCAGATGCCTCCAGCTATTAC[A/G]AGTCCCATGGCAACAAGGATCAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTNL10 PubMed Links

Gene Details

Gene
BTNL10
Gene Name
butyrophilin like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287262.1 2024 Intron NP_001274191.1
XM_011544015.2 2024 Silent Mutation CTC,CTT L221L XP_011542317.1
XM_016999986.1 2024 Silent Mutation CTC,CTT L221L XP_016855475.1
XM_016999987.1 2024 Intron XP_016855476.1

View Full Product Details