Product Details

SNP ID
rs9471065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39315101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTGGGTCTCCTGCCTGGAGC[C/T]GCCTTGGCTCCACAGCCTTGCCTCA
Phenotype
MIM: 607369 MIM: 607370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK16 PubMed Links

Gene Details

Gene
KCNK16
Gene Name
potassium two pore domain channel subfamily K member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135105.1 760 UTR 3 NP_001128577.1
NM_001135106.1 760 Intron NP_001128578.1
NM_001135107.1 760 Silent Mutation GCA,GCG A230A NP_001128579.1
NM_032115.3 760 Silent Mutation GCA,GCG A277A NP_115491.1
XM_011514935.2 760 Silent Mutation GCA,GCG A165A XP_011513237.1
XM_017011346.1 760 Silent Mutation GCA,GCG A230A XP_016866835.1
Gene
KCNK17
Gene Name
potassium two pore domain channel subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135111.1 760 Intron NP_001128583.1
NM_031460.3 760 Intron NP_113648.2

View Full Product Details