Product Details

SNP ID
rs34143060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2832114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAGAGGAGCCGACCACTGAGCC[A/T]CCAGTTAATCTCACATACTCAGCAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRSS30P PubMed Links

Gene Details

Gene
PRSS30P
Gene Name
protease, serine, 30 pseudogene
There are no transcripts associated with this gene.

Gene
ZG16B
Gene Name
zymogen granule protein 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145252.2 658 Silent Mutation CCA,CCT P194P NP_660295.2

View Full Product Details