Product Details

SNP ID
rs61733355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:193402838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAACATCTTTTAATCATCATCCA[C/T]AGGGCTCGATTTTCAATAACAGCCT
Phenotype
MIM: 609556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP13A4 PubMed Links

Gene Details

Gene
ATP13A4
Gene Name
ATPase 13A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032279.3 3364 Silent Mutation CTA,CTG L1135L NP_115655.2
XM_011513232.2 3364 Intron XP_011511534.1
XM_017007318.1 3364 Silent Mutation CTA,CTG L1026L XP_016862807.1
XM_017007319.1 3364 Intron XP_016862808.1

View Full Product Details