Product Details

SNP ID
rs117271925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22513992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCGGAACTCCTGGAAGTCAGTC[A/G]AACATATTTTCAGCTAGAATCCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF280A PubMed Links

Gene Details

Gene
ZNF280A
Gene Name
zinc finger protein 280A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080740.4 1903 UTR 3 NP_542778.2
Gene
ZNF280B
Gene Name
zinc finger protein 280B
There are no transcripts associated with this gene.

View Full Product Details