Product Details

SNP ID
rs200522940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66844830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCTGGGGTCTCACTGTCTGAC[A/G]GCCCGTCACTCACAGCTTGTCCTGA
Phenotype
MIM: 616109 MIM: 608786 MIM: 605385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf80 PubMed Links

Gene Details

Gene
C11orf80
Gene Name
chromosome 11 open reading frame 80
There are no transcripts associated with this gene.

Gene
PC
Gene Name
pyruvate carboxylase
There are no transcripts associated with this gene.

Gene
RCE1
Gene Name
Ras converting CAAX endopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032279.1 Intron NP_001027450.1
NM_005133.2 Intron NP_005124.1

View Full Product Details