Product Details

SNP ID
rs116762015
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:161365763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCCCAGTAACTGACTTAGACCT[C/T]TGTATTATTTCCTTTCTTTACCTTT
Phenotype
MIM: 616119 MIM: 602413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP126 PubMed Links

Gene Details

Gene
CFAP126
Gene Name
cilia and flagella associated protein 126
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013625.3 Intron NP_001013647.2
Gene
SDHC
Gene Name
succinate dehydrogenase complex subunit C
There are no transcripts associated with this gene.

View Full Product Details