Product Details

SNP ID
rs79796809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158804401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGATTTTTCACTCTTGTTTCTTT[C/T]GTTTTCTTTAGAGCAAGTGTAGCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD1 PubMed Links

Gene Details

Gene
MFSD1
Gene Name
major facilitator superfamily domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167903.1 Intron NP_001161375.1
NM_001289406.1 Intron NP_001276335.1
NM_001289407.1 Intron NP_001276336.1
NM_022736.2 Intron NP_073573.2
XM_006713730.2 Intron XP_006713793.1

View Full Product Details