Product Details

SNP ID
rs61736009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21523010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGCAGGCCTTTCTGCTGCTGCT[C/G]TCTCCTGGCTCCTTTCTCATTGCCT
Phenotype
MIM: 602219
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SALL2 PubMed Links

Gene Details

Gene
SALL2
Gene Name
spalt like transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291446.1 3085 Missense Mutation GAC,GAG D771E NP_001278375.1
NM_001291447.1 3085 Missense Mutation GAC,GAG D769E NP_001278376.1
NM_005407.2 3085 Missense Mutation GAC,GAG D906E NP_005398.2
XM_011537064.1 3085 Missense Mutation GAC,GAG D906E XP_011535366.1
XM_011537065.1 3085 Missense Mutation GAC,GAG D904E XP_011535367.1

View Full Product Details