Product Details

SNP ID
rs16856980
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:232399090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGTGACTTCAAAGGGACAAGGG[A/G]CATTTCCCAGTGGTCCCTTGATGAG
Phenotype
MIM: 611609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIPA1L2 PubMed Links

Gene Details

Gene
SIPA1L2
Gene Name
signal induced proliferation associated 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020808.4 5668 UTR 3 NP_065859.3
XM_005273211.2 5668 UTR 3 XP_005273268.1
XM_005273212.4 5668 UTR 3 XP_005273269.1
XM_005273213.4 5668 UTR 3 XP_005273270.1
XM_011544243.2 5668 UTR 3 XP_011542545.1
XM_017001896.1 5668 UTR 3 XP_016857385.1
XM_017001897.1 5668 UTR 3 XP_016857386.1
XM_017001898.1 5668 UTR 3 XP_016857387.1

View Full Product Details