Product Details
- SNP ID
-
rs61730956
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:100606369 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACATTTTTCTTACCCAAATTACCA[C/T]GGTAACTTGCAGCATTTGGATTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMTC4
PubMed Links
Gene Details
- Gene
- TMTC4
- Gene Name
- transmembrane and tetratricopeptide repeat containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001079669.2 |
2379 |
Missense Mutation |
CAT,CGT |
H689R |
NP_001073137.1 |
NM_001286453.1 |
2379 |
Missense Mutation |
CAT,CGT |
H578R |
NP_001273382.1 |
NM_032813.3 |
2379 |
Missense Mutation |
CAT,CGT |
H708R |
NP_116202.2 |
XM_011521121.2 |
2379 |
Missense Mutation |
CAT,CGT |
H766R |
XP_011519423.1 |
XM_011521122.2 |
2379 |
Missense Mutation |
CAT,CGT |
H766R |
XP_011519424.1 |
XM_011521123.2 |
2379 |
Missense Mutation |
CAT,CGT |
H747R |
XP_011519425.1 |
XM_011521124.2 |
2379 |
Missense Mutation |
CAT,CGT |
H747R |
XP_011519426.1 |
XM_011521125.1 |
2379 |
Intron |
|
|
XP_011519427.1 |
XM_017020796.1 |
2379 |
Missense Mutation |
CAT,CGT |
H689R |
XP_016876285.1 |
XM_017020797.1 |
2379 |
Missense Mutation |
CAT,CGT |
H530R |
XP_016876286.1 |
XM_017020798.1 |
2379 |
Missense Mutation |
CAT,CGT |
H472R |
XP_016876287.1 |
XM_017020799.1 |
2379 |
Missense Mutation |
CAT,CGT |
H472R |
XP_016876288.1 |
XM_017020800.1 |
2379 |
Intron |
|
|
XP_016876289.1 |
View Full Product Details