Product Details

SNP ID
rs36054910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53253567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGGAGGCCTGCGCTGCCTCCT[G/T]TCGGACCGCCGCGTGCTGCTGTTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MFSD5 PubMed Links

Gene Details

Gene
MFSD5
Gene Name
major facilitator superfamily domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170790.1 877 Silent Mutation CTG,CTT L351L NP_001164261.1
NM_032889.4 877 Silent Mutation CTG,CTT L244L NP_116278.3
XM_005269197.1 877 Silent Mutation CTG,CTT L351L XP_005269254.1
XM_005269198.4 877 Silent Mutation CTG,CTT L244L XP_005269255.1

View Full Product Details