Product Details

SNP ID
rs7077672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70774154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGGCTCCCTTCTCCAGGTCCCA[C/T]TGGCCTCACCTGGGAGGACAGAAGG
Phenotype
MIM: 612640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBATA PubMed Links

Gene Details

Gene
TBATA
Gene Name
thymus, brain and testes associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318241.1 Intron NP_001305170.1
NM_001318242.1 Intron NP_001305171.1
NM_001318243.1 Intron NP_001305172.1
NM_152710.3 Intron NP_689923.3
XM_017015845.1 Intron XP_016871334.1
XM_017015846.1 Intron XP_016871335.1
XM_017015847.1 Intron XP_016871336.1
XM_017015848.1 Intron XP_016871337.1
XM_017015849.1 Intron XP_016871338.1
XM_017015850.1 Intron XP_016871339.1
XM_017015851.1 Intron XP_016871340.1
XM_017015852.1 Intron XP_016871341.1
XM_017015853.1 Intron XP_016871342.1
XM_017015854.1 Intron XP_016871343.1
XM_017015855.1 Intron XP_016871344.1
XM_017015856.1 Intron XP_016871345.1
XM_017015857.1 Intron XP_016871346.1
XM_017015858.1 Intron XP_016871347.1
XM_017015859.1 Intron XP_016871348.1
XM_017015860.1 Intron XP_016871349.1
XM_017015861.1 Intron XP_016871350.1
XM_017015862.1 Intron XP_016871351.1

View Full Product Details