Product Details

SNP ID
rs73272359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73499319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATACATACAAGGGAAAGAATGAA[C/T]CAAGCTGTGCTTTTGATTCCATTTC
Phenotype
MIM: 114106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP3CB PubMed Links

Gene Details

Gene
PPP3CB
Gene Name
protein phosphatase 3 catalytic subunit beta
There are no transcripts associated with this gene.

Gene
PPP3CB-AS1
Gene Name
PPP3CB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
USP54
Gene Name
ubiquitin specific peptidase 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320437.1 Intron NP_001307366.1
NM_001320441.1 Intron NP_001307370.1
NM_152586.3 Intron NP_689799.3
XM_005269582.2 Intron XP_005269639.1
XM_011539368.2 Intron XP_011537670.1
XM_017015759.1 Intron XP_016871248.1
XM_017015760.1 Intron XP_016871249.1
XM_017015761.1 Intron XP_016871250.1
XM_017015762.1 Intron XP_016871251.1
XM_017015763.1 Intron XP_016871252.1
XM_017015764.1 Intron XP_016871253.1
XM_017015765.1 Intron XP_016871254.1
XM_017015766.1 Intron XP_016871255.1
XM_017015767.1 Intron XP_016871256.1
XM_017015768.1 Intron XP_016871257.1
XM_017015769.1 Intron XP_016871258.1
XM_017015770.1 Intron XP_016871259.1
XM_017015771.1 Intron XP_016871260.1
XM_017015772.1 Intron XP_016871261.1
XM_017015773.1 Intron XP_016871262.1
XM_017015774.1 Intron XP_016871263.1
XM_017015775.1 Intron XP_016871264.1
XM_017015776.1 Intron XP_016871265.1
XM_017015777.1 Intron XP_016871266.1
XM_017015778.1 Intron XP_016871267.1
XM_017015779.1 Intron XP_016871268.1
XM_017015780.1 Intron XP_016871269.1
XM_017015781.1 Intron XP_016871270.1
XM_017015782.1 Intron XP_016871271.1
XM_017015783.1 Intron XP_016871272.1
XM_017015784.1 Intron XP_016871273.1
XM_017015785.1 Intron XP_016871274.1
XM_017015786.1 Intron XP_016871275.1

View Full Product Details