Product Details
- SNP ID
-
rs3773305
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:12837054 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATGAGTCCACTCAAATTCAAAGCAA[C/T]TGTATTTTTAAATGTTTCTACGTTT
- Phenotype
-
MIM: 610403
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAND2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80067234] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAND2
- Gene Name
- cullin associated and neddylation dissociated 2 (putative)
There are no transcripts associated with this gene.
- Gene
- RPL32
- Gene Name
- ribosomal protein L32
- Gene
- SNORA7A
- Gene Name
- small nucleolar RNA, H/ACA box 7A
There are no transcripts associated with this gene.
View Full Product Details