Product Details

SNP ID
rs3788409
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:28802163 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTAAGTCGGATTAGCTGGTCAATT[G/T]TATTTTGGTAGGGAGGGAACAGGAT
Phenotype
MIM: 194355 MIM: 612062
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
XBP1 PubMed Links

Gene Details

Gene
XBP1
Gene Name
X-box binding protein 1
There are no transcripts associated with this gene.

Gene
ZNRF3
Gene Name
zinc and ring finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206998.1 Intron NP_001193927.1
NM_032173.3 Intron NP_115549.2
XM_011530435.2 Intron XP_011528737.1
XM_011530436.2 Intron XP_011528738.1
XM_011530437.1 Intron XP_011528739.1
XM_011530438.2 Intron XP_011528740.1
XM_017028990.1 Intron XP_016884479.1

View Full Product Details