Product Details
- SNP ID
-
rs3213990
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43378028 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGTTTTTATTTATTCATTCATTTAA[A/C]AATATCTGCTTGGTTATCATGTTAA
- Phenotype
-
MIM: 609610
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TUBGCP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3213991] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TUBGCP4
- Gene Name
- tubulin gamma complex associated protein 4
- Gene
- ZSCAN29
- Gene Name
- zinc finger and SCAN domain containing 29
There are no transcripts associated with this gene.
View Full Product Details