Product Details
- SNP ID
-
rs2450135
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:78216949 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATTGGAACACCAAGGCATGGCCCCT[A/G]CACTTCTGAGTGGCTGGACCCATAT
- Phenotype
-
MIM: 606203
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GAB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3740677] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAB2
- Gene Name
- GRB2 associated binding protein 2
- Gene
- USP35
- Gene Name
- ubiquitin specific peptidase 35
View Full Product Details