Product Details
- SNP ID
-
rs3752183
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56601810 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGAATGGCCAAACTAGCTCTCTGGA[C/T]TCTTAGGAGACACAGGTGGGCACCT
- Phenotype
-
MIM: 194545
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF71
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs45498492] are located under a probe and SNP(s) [rs73934487] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF71
- Gene Name
- zinc finger protein 71
View Full Product Details