Product Details
- SNP ID
-
rs3796490
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:76307612 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCTTTCAGCACTGTCGGCAGTCCCC[T/C]CCACGCTGCCCCAGAATAGGGCCGG
- Phenotype
-
MIM: 607406
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC158
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3796489] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC158
- Gene Name
- coiled-coil domain containing 158
There are no transcripts associated with this gene.
- Gene
- FAM47E-STBD1
- Gene Name
- FAM47E-STBD1 readthrough
- Gene
- STBD1
- Gene Name
- starch binding domain 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003943.4 |
|
Intron |
|
|
NP_003934.1 |
View Full Product Details