Product Details

SNP ID
rs1721100
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:16992989 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATAGACTTTAATATTTTGAACGTCT[G/C]CTTGGGTCATTATTTTATGATGGGA
Phenotype
MIM: 605558
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
FGF20 PubMed Links

Gene Details

Gene
FGF20
Gene Name
fibroblast growth factor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019851.2 852 UTR 3 NP_062825.1

View Full Product Details