Product Details

SNP ID
hCV25921946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173232870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCTAGGCATGGCTTGCCATCG[C/T]GCACCAGCACTGGCACCGCGATCCT
Phenotype
MIM: 600584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKX2-5 PubMed Links

Gene Details

Gene
NKX2-5
Gene Name
NK2 homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166175.1 1158 UTR 3 NP_001159647.1
NM_001166176.1 1158 UTR 3 NP_001159648.1
NM_004387.3 1158 Missense Mutation CAC,CGC H225R NP_004378.1

View Full Product Details