Product Details

SNP ID
rs73372042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5290854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCAAAGTGTCCCTGTCCCACC[A/G]CCTCTAGCTACAGGCTATCGTCTCC
Phenotype
MIM: 602126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB14 PubMed Links

Gene Details

Gene
ZBTB14
Gene Name
zinc finger and BTB domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143823.2 1516 UTR 3 NP_001137295.1
NM_001243702.1 1516 UTR 3 NP_001230631.1
NM_001243704.1 1516 UTR 3 NP_001230633.1
NM_003409.4 1516 UTR 3 NP_003400.2

View Full Product Details