Product Details

SNP ID
rs75310182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22244882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGAGGGAACCCCCAGATCCCA[A/C]AGACTCCTGCCCCTTCCTTCATCCT
Phenotype
MIM: 605141
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VPREB1 PubMed Links

Gene Details

Gene
VPREB1
Gene Name
pre-B lymphocyte 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303509.1 Intron NP_001290438.1
NM_007128.3 Intron NP_009059.1

View Full Product Details