Product Details
- SNP ID
-
rs34901659
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:53028921 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATTTGGGGGATGTGGGACCTCCA[G/A]TTCCCAGCCCCGGCTTCAGCTCTTT
- Phenotype
-
MIM: 614061
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OLFM4
PubMed Links
Gene Details
- Gene
- OLFM4
- Gene Name
- olfactomedin 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006418.4 |
181 |
Missense Mutation |
ATT,GTT |
I29V |
NP_006409.3 |
View Full Product Details