Product Details

SNP ID
rs61748993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153905157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCAGCCCCACCATGCTCATGGC[A/G]TCCACCACTTCCGGTAAGGCTTGCC
Phenotype
MIM: 300023 MIM: 300538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP4 PubMed Links

Gene Details

Gene
ARHGAP4
Gene Name
Rho GTPase activating protein 4
There are no transcripts associated with this gene.

Gene
AVPR2
Gene Name
arginine vasopressin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000054.4 83 Silent Mutation GCA,GCG A4A NP_000045.1
NM_001146151.1 83 Silent Mutation GCA,GCG A4A NP_001139623.1
XM_006724828.3 83 Silent Mutation GCA,GCG A4A XP_006724891.1

View Full Product Details