Product Details

SNP ID
rs12941739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55751132 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTGGCCGCCGGAAGCTTCTCGG[A/C]GGAGCAGTTCTGGGAGGCCTGCGCC
Phenotype
MIM: 606055
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCTP PubMed Links

Gene Details

Gene
PCTP
Gene Name
phosphatidylcholine transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102402.2 3 UTR 5 NP_001095872.1
NM_021213.3 3 Missense Mutation GAG,GCG E10A NP_067036.2
XM_005257549.1 3 Missense Mutation GAG,GCG E10A XP_005257606.1
XM_005257550.3 3 Missense Mutation GAG,GCG E10A XP_005257607.1
XM_017024913.1 3 UTR 5 XP_016880402.1

View Full Product Details