Product Details

SNP ID
rs972551638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55751088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCTGCCCTCCAGGCGGAGGAG[C/T]CCGGACTGCGGAAGGATGGAGCTGG
Phenotype
MIM: 606055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCTP PubMed Links

Gene Details

Gene
PCTP
Gene Name
phosphatidylcholine transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102402.2 110 Intron NP_001095872.1
NM_021213.3 110 UTR 5 NP_067036.2

View Full Product Details