Product Details

SNP ID
rs6878145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141338985 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAGGAGGGAAATGGCGGCTCTGC[A/G]AAAGTTGCCACACTGCAGAAAGCTG
Phenotype
MIM: 606288 MIM: 606289 MIM: 606290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 226 Intron NP_061735.1
NM_031993.1 226 Intron NP_114382.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018915.3 226 Missense Mutation CAA,CGA Q5R NP_061738.1
NM_032009.2 226 Missense Mutation CAA,CGA Q5R NP_114398.1
Gene
PCDHGA3
Gene Name
protocadherin gamma subfamily A, 3
There are no transcripts associated with this gene.

View Full Product Details