Product Details

SNP ID
rs143532876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56511588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAATTTAGAGGAAGGCTTTAGC[A/T]CTCACTCCTATAATCTCCTGTAATC
Phenotype
MIM: 612405 MIM: 602122
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL9 PubMed Links

Gene Details

Gene
ARL9
Gene Name
ADP ribosylation factor like GTPase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206919.1 Intron NP_996802.1
XM_005265719.3 Intron XP_005265776.1
XM_005265720.3 Intron XP_005265777.1
XM_006714036.3 Intron XP_006714099.1
Gene
SRP72
Gene Name
signal recognition particle 72
There are no transcripts associated with this gene.

View Full Product Details