Product Details

SNP ID
rs143183943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38568726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAGGATTTATCTAGTACTTAAG[C/G]TAGGGAGTAGAGTTCCACAAAATCA
Phenotype
MIM: 611208
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPLL PubMed Links

Gene Details

Gene
HNRNPLL
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142650.1 Intron NP_001136122.1
NM_138394.3 Intron NP_612403.2
XM_005264639.3 Intron XP_005264696.2
XM_005264640.3 Intron XP_005264697.2
XM_011533165.2 Intron XP_011531467.1
XM_011533166.2 Intron XP_011531468.1

View Full Product Details