Product Details

SNP ID
rs112790429
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:211481102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGCCTGCAGCCCAGCAAGGGT[C/T]CCCATCCCAGTGCTCACCTGAGGAT
Phenotype
MIM: 180040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RD3 PubMed Links

Gene Details

Gene
RD3
Gene Name
retinal degeneration 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164688.1 Intron NP_001158160.1
NM_183059.2 Intron NP_898882.1
XM_017001151.1 Intron XP_016856640.1

View Full Product Details