Product Details
- SNP ID
-
rs61730378
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20510810 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAACAAAGACTATCTTAGGCTTGA[C/G]CAGACAGATAGAGAATGCAATGATA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RNASE10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75463308] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RNASE10
- Gene Name
- ribonuclease A family member 10 (inactive)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012975.1 |
339 |
Missense Mutation |
GAC,GAG |
D113E |
NP_001012993.1 |
View Full Product Details