Product Details

SNP ID
rs11118237
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:207645374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGCGCGTCCGCGGCGAGGCTAGA[A/G]CTCTGCTCAGTCAGTCGGGCAGGAG
Phenotype
MIM: 120620 MIM: 605886
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CR1 PubMed Links

Gene Details

Gene
CR1
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
There are no transcripts associated with this gene.

Gene
CR1L
Gene Name
complement component 3b/4b receptor 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175710.1 Intron NP_783641.1

View Full Product Details