Product Details

SNP ID
hCV25924820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73683272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGAGGGGACGCCGAGCAGATCA[C/T]ACAGCGCCGTGCGCGAATACGAGCA
Phenotype
MIM: 615733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC30 PubMed Links

Gene Details

Gene
DNAJC30
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
There are no transcripts associated with this gene.

Gene
WBSCR22
Gene Name
Williams-Beuren syndrome chromosome region 22
There are no transcripts associated with this gene.

View Full Product Details