Product Details

SNP ID
rs78223075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113005387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGACACAATGCTACTTTAAATG[G/T]AGACTGAACTGAAATGAACTCCTGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GTPBP8 PubMed Links

Gene Details

Gene
GTPBP8
Gene Name
GTP binding protein 8 (putative)
There are no transcripts associated with this gene.

Gene
NEPRO
Gene Name
nucleolus and neural progenitor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319109.1 1640 UTR 3 NP_001306038.1
NM_001319110.1 1640 UTR 3 NP_001306039.1
NM_001319111.1 1640 UTR 3 NP_001306040.1
NM_001319112.1 1640 UTR 3 NP_001306041.1
NM_001319114.1 1640 UTR 3 NP_001306043.1
NM_001319115.1 1640 UTR 3 NP_001306044.1
NM_015412.3 1640 UTR 3 NP_056227.2
XM_005247276.2 1640 UTR 3 XP_005247333.1
XM_011512635.2 1640 UTR 3 XP_011510937.1
XM_011512636.2 1640 UTR 3 XP_011510938.1
XM_011512638.2 1640 UTR 3 XP_011510940.1
XM_017006102.1 1640 UTR 3 XP_016861591.1
XM_017006103.1 1640 Intron XP_016861592.1

View Full Product Details