Product Details

SNP ID
rs115422575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143273179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTCCTCACCGCCCCCTCTGTAG[G/T]AGCATGGCGCTGGAGCCTGCCCTGT
Phenotype
MIM: 165280
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLI4 PubMed Links

Gene Details

Gene
GLI4
Gene Name
GLI family zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138465.3 Intron NP_612474.1
Gene
MINCR
Gene Name
MYC-induced long noncoding RNA
There are no transcripts associated with this gene.

View Full Product Details