Product Details

SNP ID
rs33955038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1318376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCAGCCCGGGCGGCCTTCAGTCC[A/G]TGTGGGGCGCCCGCGTGGCCTTCTC
Phenotype
MIM: 612585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPTM1L PubMed Links

Gene Details

Gene
CLPTM1L
Gene Name
CLPTM1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030782.4 1803 Missense Mutation NP_110409.2
XM_011514144.2 1803 Missense Mutation XP_011512446.1
Gene
MIR4457
Gene Name
microRNA 4457
There are no transcripts associated with this gene.

View Full Product Details