Product Details

SNP ID
rs149050888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15675171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTAGCCCTTACGTGAAAGGCGT[G/T]GCGTAGTCGCAAGGGTGTAAAAAAG
Phenotype
MIM: 300631
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CA5BP1 PubMed Links

Gene Details

Gene
CA5BP1
Gene Name
carbonic anhydrase 5B pseudogene 1
There are no transcripts associated with this gene.

Gene
TMEM27
Gene Name
transmembrane protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020665.5 Intron NP_065716.1
XM_017029680.1 Intron XP_016885169.1
XM_017029681.1 Intron XP_016885170.1

View Full Product Details