Product Details

SNP ID
rs41288827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27364399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTATTAACCCTGTGTTTCCCCC[A/G]TCACTGGTCACTGCTCTTGACTCGT
Phenotype
MIM: 606687 MIM: 604883 MIM: 605963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B4 PubMed Links

Gene Details

Gene
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034116.1 1713 UTR 3 NP_001029288.1
NM_001318965.1 1713 UTR 3 NP_001305894.1
NM_001318966.1 1713 UTR 3 NP_001305895.1
NM_001318967.1 1713 UTR 3 NP_001305896.1
NM_001318968.1 1713 UTR 3 NP_001305897.1
NM_001318969.1 1713 UTR 3 NP_001305898.1
NM_015636.3 1713 UTR 3 NP_056451.3
NM_172195.3 1713 UTR 3 NP_751945.2
XM_006712132.1 1713 UTR 3 XP_006712195.1
XM_011533147.2 1713 UTR 3 XP_011531449.1
Gene
GTF3C2
Gene Name
general transcription factor IIIC subunit 2
There are no transcripts associated with this gene.

Gene
LOC105374363
Gene Name
uncharacterized LOC105374363
There are no transcripts associated with this gene.

Gene
SNX17
Gene Name
sorting nexin 17
There are no transcripts associated with this gene.

View Full Product Details