Product Details

SNP ID
rs35773045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19072092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTCCTGGGTCGGACCAGCGTC[T/G]GGCCAGTCATTCAGGTGGGTGCTTC
Phenotype
MIM: 601683
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
COQ7 PubMed Links

Gene Details

Gene
COQ7
Gene Name
coenzyme Q7, hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190983.1 257 Missense Mutation GGG,TGG G42W NP_001177912.1
NM_016138.4 257 Missense Mutation GGG,TGG G80W NP_057222.2
Gene
LOC102723385
Gene Name
uncharacterized LOC102723385
There are no transcripts associated with this gene.

Gene
TMC7
Gene Name
transmembrane channel like 7
There are no transcripts associated with this gene.

View Full Product Details