Product Details

SNP ID
rs185400827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57758620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGAGAGGCCCACCCTCCTCCAC[A/G]CTCCCGCCCTCCTCACGGGGCCCAG
Phenotype
MIM: 602703 MIM: 604535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KATNB1 PubMed Links

Gene Details

Gene
KATNB1
Gene Name
katanin regulatory subunit B1
There are no transcripts associated with this gene.

Gene
KIFC3
Gene Name
kinesin family member C3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130099.1 3519 UTR 3 NP_001123571.1
NM_001130100.1 3519 UTR 3 NP_001123572.1
NM_001318710.1 3519 UTR 3 NP_001305639.1
NM_001318711.1 3519 UTR 3 NP_001305640.1
NM_001318712.1 3519 UTR 3 NP_001305641.1
NM_001318713.1 3519 UTR 3 NP_001305642.1
NM_001318714.1 3519 UTR 3 NP_001305643.1
NM_001318715.1 3519 UTR 3 NP_001305644.1
NM_005550.3 3519 UTR 3 NP_005541.3
XM_005255937.1 3519 UTR 3 XP_005255994.1
XM_005255944.1 3519 UTR 3 XP_005256001.1
XM_006721188.1 3519 UTR 3 XP_006721251.1
XM_011523075.1 3519 UTR 3 XP_011521377.1
XM_011523076.1 3519 UTR 3 XP_011521378.1
XM_011523077.1 3519 UTR 3 XP_011521379.1
XM_011523078.1 3519 UTR 3 XP_011521380.1
XM_011523079.1 3519 UTR 3 XP_011521381.1
XM_017023219.1 3519 UTR 3 XP_016878708.1
XM_017023220.1 3519 UTR 3 XP_016878709.1
XM_017023221.1 3519 UTR 3 XP_016878710.1
XM_017023222.1 3519 UTR 3 XP_016878711.1
XM_017023223.1 3519 UTR 3 XP_016878712.1
XM_017023224.1 3519 UTR 3 XP_016878713.1
XM_017023225.1 3519 UTR 3 XP_016878714.1
XM_017023226.1 3519 UTR 3 XP_016878715.1
XM_017023227.1 3519 UTR 3 XP_016878716.1

View Full Product Details