Product Details

SNP ID
rs61739070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36754830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAAAGGCCTGCAGCTGTTTCAC[C/T]GATCCTTGCAGTTCATCCATCTGTA
Phenotype
MIM: 609175 MIM: 605273
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSN1 PubMed Links

Gene Details

Gene
DSN1
Gene Name
DSN1 homolog, MIS12 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145315.1 1249 Silent Mutation TCA,TCG S298S NP_001138787.1
NM_001145316.1 1249 Silent Mutation TCA,TCG S298S NP_001138788.1
NM_001145317.1 1249 Silent Mutation TCA,TCG S191S NP_001138789.1
NM_001145318.1 1249 Silent Mutation TCA,TCG S282S NP_001138790.1
NM_024918.3 1249 Silent Mutation TCA,TCG S298S NP_079194.3
XM_006723876.2 1249 Silent Mutation TCA,TCG S191S XP_006723939.1
XM_017028070.1 1249 Silent Mutation TCA,TCG S286S XP_016883559.1
XM_017028071.1 1249 Silent Mutation TCA,TCG S286S XP_016883560.1
XM_017028072.1 1249 Silent Mutation TCA,TCG S270S XP_016883561.1
Gene
NDRG3
Gene Name
NDRG family member 3
There are no transcripts associated with this gene.

View Full Product Details