Product Details

SNP ID
rs61733475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2420114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAACGTTTATCAAACACTTAAA[C/T]AGGTACTGTCCGGCCACTCCTGGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC284009 PubMed Links

Gene Details

Gene
LOC284009
Gene Name
uncharacterized LOC284009
There are no transcripts associated with this gene.

Gene
METTL16
Gene Name
methyltransferase like 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024086.3 1736 Silent Mutation CTA,CTG L515L NP_076991.3
XM_005256801.2 1736 Silent Mutation CTA,CTG L297L XP_005256858.1
XM_017025045.1 1736 Silent Mutation CTA,CTG L297L XP_016880534.1
XM_017025046.1 1736 Silent Mutation CTA,CTG L297L XP_016880535.1

View Full Product Details