Product Details
- SNP ID
-
rs34656422
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82392460 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAGCTGAAGGCGATGGTGATGG[C/T]GTAACGGGTGCCCCAGTGGACCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105376790
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11903] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105376790
- Gene Name
- uncharacterized LOC105376790
There are no transcripts associated with this gene.
- Gene
- OGFOD3
- Gene Name
- 2-oxoglutarate and iron dependent oxygenase domain containing 3
View Full Product Details