Product Details

SNP ID
rs111226417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:98874188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTACAAATGTCTTAATTCTTAC[C/G]ATTAAAACAACTAAAAACAAAACCC
Phenotype
MIM: 605654
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXL4 PubMed Links

Gene Details

Gene
FBXL4
Gene Name
F-box and leucine rich repeat protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278716.1 2381 UTR 3 NP_001265645.1
NM_012160.4 2381 UTR 3 NP_036292.2
XM_005266930.2 2381 UTR 3 XP_005266987.1
XM_011535748.2 2381 Intron XP_011534050.1
XM_017010726.1 2381 UTR 3 XP_016866215.1
XM_017010727.1 2381 UTR 3 XP_016866216.1
XM_017010728.1 2381 UTR 3 XP_016866217.1

View Full Product Details